Ethylammonium chloride

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Aminoethane hydrochloride, Ethylamine hydrochloride

9745.1
ROTH9745.1EA 0 GBP
ROTH9745.1
Ethylammonium chloride
Ethylammonium chloride
Formula: C₂H₈ClN
MDL Number: MFCD00012885
CAS Number: 557-66-4
EINECS: 209-182-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR