Dihydrojasmone

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

8735.1
ROTH8735.1EA 0 GBP
ROTH8735.1
Dihydrojasmone
Dihydrojasmone
Formula: C₁₁H₁₈O
MDL Number: MFCD00036480
CAS Number: 1128-08-1
EINECS: 214-434-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR