Terpineol, mixture of isomers

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

8672.1 8672.2
ROTH8672.1EA 0 GBP
ROTH8672.1 ROTH8672.2
Terpineol, mixture of isomers
Terpineol, mixture of isomers
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
MDL Number: MFCD00166983
CAS Number: 8000-41-7
EINECS: 232-268-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR