Copper(II) sulphate pentahydrate

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

8175.1 8175.2 8175.5
ROTH8175.1EA 0 GBP
ROTH8175.1 ROTH8175.2 ROTH8175.5
Copper(II) sulphate pentahydrate
Copper (II) sulphate pentahydrate
Formula: CuSO₄·5H₂O
Density: 2.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
UN: 3077
ADR: 9,III
REACH: 01-2119520566-40
Merck Index: 13,02682

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR