Guaiacol

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

7308.1
ROTH7308.1EA 0 GBP
ROTH7308.1
Guaiacol
Guaiacol
Formula: C₇H₈O₂
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 28…32 °C
Density: 1.1534 g/cm³ (0 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00002185
CAS Number: 90-05-1
EINECS: 201-964-7
UN: 2810
ADR: 6.1,III
Merck Index: 13,04568

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR