MOPSO (3-(4-morpholino)-2-hydroxypropanesulphonic acid)

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-Hydroxy-3-morpholinopropane-1-sulphonic acid

7117.1 7117.2 7117.3
ROTH7117.1EA 0 GBP
ROTH7117.1 ROTH7117.2 ROTH7117.3
MOPSO (3-(4-morpholino)-2-hydroxypropanesulphonic acid)
MOPSO (3-(4-Morpholino)-2-hydroxypropanesulphonic acid)
Formula: C₇H₁₅NO₅S
MW: 225.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009629
CAS Number: 68399-77-9
EINECS: 269-989-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR