Corn oil

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

6212.1
ROTH6212.1EA 0 GBP
ROTH6212.1
Corn oil
Corn oil
Melting Pt: –10 °C
Density: 0.918 g/cm³ (20 °C)
MDL Number: MFCD00130865
CAS Number: 8001-30-7
EINECS: 232-281-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR