Starch, soluble

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Starch, Vitex® iodine indicator, Amylodextrin

4701.1 4701.2
ROTH4701.1EA 0 GBP
ROTH4701.1 ROTH4701.2
Starch, soluble
Starch, soluble
Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-84-9
EINECS: 232-686-4
Merck Index: 13,08878

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR