Ammonium thiocyanate

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

4477.1 4477.2 4477.3 4477.4
ROTH4477.1EA 0 GBP
ROTH4477.1 ROTH4477.2 ROTH4477.3 ROTH4477.4
Ammonium thiocyanate
Ammonium thiocyanate
Formula: NH₄SCN
Melting Pt: 150 °C
Density: 1.305 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011428
CAS Number: 1762-95-4
EINECS: 217-175-6
Merck Index: 13,00564

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR