5-Fluoroorotic acid

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

4015.3
ROTH4015.3EA 0 GBP
ROTH4015.3
5-Fluoroorotic acid
5-Fluoroorotic acid
Formula: C₅H₃FN₂O₄
MDL Number: MFCD00042526
CAS Number: 703-95-7
EINECS: 211-876-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR