β-Ionone

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (E)-4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one, (E)-4-(2,6,6-Trimethylcyclohex-1-en-1-yl)but-3-en-2-one, beta-Ionone

3713.1
ROTH3713.1EA 0 GBP
ROTH3713.1
β-Ionone
β-Ionone
Formula: C₁₃H₂₀O
MW: 192.30 g/mol
MDL Number: MFCD00001549
CAS Number: 79-77-6
EINECS: 201-224-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR