Evening primrose oil

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

3691.1 3691.2
ROTH3691.1EA 0 GBP
ROTH3691.1 ROTH3691.2
Evening primrose oil
Evening primrose oil
MDL Number: MFCD00131042
CAS Number: 90028-66-3
EINECS: 289-859-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR