N-Bromosuccinimide

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: NBS, 1-Bromopyrrolidine-2,5-dione

3261.1 3261.2
ROTH3261.1EA 0 GBP
ROTH3261.1 ROTH3261.2
N-Bromosuccinimide
N-Bromosuccinimide
Formula: C₄H₄BrNO₂
MW: 177.99 g/mol
Boiling Pt: 339
Melting Pt: 174…179 °C
Density: 2.098 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00005510
CAS Number: 128-08-5
EINECS: 204-877-2
UN: 3261
ADR: 8 III
Merck Index: 13,01423

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR