Aluminium carbide

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Aluminum carbide, Tetraaluminium tricarbide

3094.1 3094.2
ROTH3094.1EA 0 GBP
ROTH3094.1 ROTH3094.2
Aluminium carbide
Aluminium carbide
Formula: C₃Al₄
Melting Pt: 2100 °C
Density: 2.36 g/cm³ (20 °C)
MDL Number: MFCD00003425
CAS Number: 1299-86-1
EINECS: 215-076-2
UN: 1394
ADR: 4.3,II
Merck Index: 12,00345

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR