Base materials for microbiology media
Supplier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2363.2 5210.2 8952.2 2363.1 2363.3 2365.1 2365.2 2365.3 2366.1 2366.2 2366.3 A157.1 A157.2 AE41.1 8952.1 8952.3 8986.1 8986.2 8986.3 2363.5 8986.4
ROTH2363.2EA 0 GBP
ROTH2363.2 ROTH5210.2 ROTH8952.2 ROTH2363.1 ROTH2363.3 ROTH2365.1 ROTH2365.2 ROTH2365.3 ROTH2366.1 ROTH2366.2 ROTH2366.3 ROTHA157.1 ROTHA157.2 ROTHAE41.1 ROTH8952.1 ROTH8952.3 ROTH8986.1 ROTH8986.2 ROTH8986.3 ROTH2363.5 ROTH8986.4
Base materials for microbiology media
Media for Microbiology Base Materials for Microbiology
Peptone from soya beans is gained from papainic digestion of soya bean flour. Peptone tryptic digest is produced by proteolytic digestion of casein with trypsin-rich pancreas extract.
  • Granulated
  • Used for culture media, for fermentation and for the production of vaccines
  • Additive to nutrient media in the production of toxins, vaccins, enzymes and further biological products
  • Low-cost peptone