Chlorotrimethylsilane

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: TMCS, TMSC, Trimethylchlorosilane

2338.1
ROTH2338.1EA 0 GBP
ROTH2338.1
Chlorotrimethylsilane
Chlorotrimethylsilane
Formula: C₃H₉ClSi
MW: 108.64 g/mol
Boiling Pt: 59
Melting Pt: –58 °C
Density: 0.8576 g/cm³ (20 °C)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00000502
CAS Number: 75-77-4
EINECS: 200-900-5
UN: 1298
REACH: 01-2119457596-25

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR