NPP ELISA alkaline phosphatase substrate

Supplier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

NPP-100 NPP-10 NPP-B500
ROCKNPP-100EA 203 GBP
ROCKNPP-100 ROCKNPP-10 ROCKNPP-B500
NPP ELISA alkaline phosphatase substrate
Electrophoresis Reagents Western Blotting Reagents

NPP ELISA alcaline phosphatase substrate is a chromogenic substrate used in immunosorbent assays.


Phosphatase-conjugated detection antibodies can act upon the pNPP to convert it to a yellow para-nitrophenol. This yellow product can be measured on a spectrophotometer at 405 nm.


Ordering information: Diluent buffer NPP-B500 for NPP-10 has to be ordered separately.


Caution: Both the substrate and buffer have to be stored at 4 °C prior to opening and should be protected from moisture and light.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR