Gas washing bottles, with filter candle, VitraPOR®
Supplier: ROBU GLASFILTER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
42093 42090 42092 42094 42130 42131 42132 42133 42134 42091
ROBU42093EA 112 GBP
ROBU42093 ROBU42090 ROBU42092 ROBU42094 ROBU42130 ROBU42131 ROBU42132 ROBU42133 ROBU42134 ROBU42091
Gas washing bottles, with filter candle, VitraPOR®
Bottles Gas Washing Bottles
They are made of borosilicate glass 3.3 and the versatile and recyclable material offers good properties for the most diverse fields.
  • Resistant to chemicals such as solvents, acids and alkalis
  • Very good temperature resistance
  • With screw cap and silicone washer