PEEK tubing, Rheodyne®
Supplier: IDEX RHEODYNE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1532
RHEO1532EA 54.9 GBP
RHEO1532
PEEK tubing, Rheodyne®
Tubing Chromatography Tubing
Polymer tubing, coloured.
  • Biocompatible, inert and easy to cut
  • Great for high pressure applications
  • Many sizes colour-coded for easier inner diameter identification
PEEK (polyetheretherketone) tubing is biocompatible, chemically inert and can be used in place of stainless steel tubing in most liquid analytical systems. Additionally, when the Ø is smaller than ¹/₁₆", PEEK tubing can be used in place of more traditional fused silica capillary tubing. PEEK tubing can be used with stainless steel nuts and ferrules, flangeless fittings, Super Flangeless™More Product Information