Revco® TDE ULT Upright Freezers, –80 °C
Supplier: REVCO TECHNOLOGIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TDE30086FVULTS TDE40086FVULTS TDE50086FVULTS TDE60086FVULTS
REVCTDE30086FVULTSEA 0 GBP
REVCTDE30086FVULTS REVCTDE40086FVULTS REVCTDE50086FVULTS REVCTDE60086FVULTS
Revco® TDE ULT Upright Freezers, –80 °C
Freezers Ultra Low Temperature Freezers
Thermo Scientific™ TDE Series –80 °C upright ULT freezers, powered by the H-Drive system, are designed for efficiency. As a result, the series enables a foundational ULT experience with the reliability and performance needed to protect your work - and the sustainability to meet global compliance.