HPLC columns, Pinnacle® DB PFP Propyl
Supplier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9419311 9419312 9419313 9419315 9419331 9419333 9419335 9419351 9419353 9419361 9419363 9419522 9419532 9419535 9419555 9419312-700 9419313-700 9419315-700 941931E 9419332-700 9419333-700 9419335-700 941933E 9419352-700 9419353-700 9419355-700 941935E 9419362-700 9419363-700 9419365-700 941936E 9419512-700 9419513-700 9419515-700 941951E 9419522-700 9419523-700 9419525-700 941952E 9419532-700 9419533-700 9419535-700 941953E 9419552-700 9419553-700 9419555-700 941955E 9419562-700 9419563-700 9419565-700 941956E 9419572-700 9419573-700 9419575-700 941957E
REST9419311EA 0 GBP
REST9419311 REST9419312 REST9419313 REST9419315 REST9419331 REST9419333 REST9419335 REST9419351 REST9419353 REST9419361 REST9419363 REST9419522 REST9419532 REST9419535 REST9419555 REST9419312-700 REST9419313-700 REST9419315-700 REST941931E REST9419332-700 REST9419333-700 REST9419335-700 REST941933E REST9419352-700 REST9419353-700 REST9419355-700 REST941935E REST9419362-700 REST9419363-700 REST9419365-700 REST941936E REST9419512-700 REST9419513-700 REST9419515-700 REST941951E REST9419522-700 REST9419523-700 REST9419525-700 REST941952E REST9419532-700 REST9419533-700 REST9419535-700 REST941953E REST9419552-700 REST9419553-700 REST9419555-700 REST941955E REST9419562-700 REST9419563-700 REST9419565-700 REST941956E REST9419572-700 REST9419573-700 REST9419575-700 REST941957E
HPLC columns, Pinnacle® DB PFP Propyl
Chromatography Columns
Pinnacle® DB PFP Propyl columns provides great retention and selectivity of charged bases, electronegative compounds, and amine-containing compounds.
  • These columns are more amenable to LC-MS because it is more reliable and effi cient with acidic mobile phases
  • They are compatible with highly aqueous mobile phases and HILIC separations