GC columns, Rt-XLSulfur (packed and micropacked)
Supplier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
19804 19805 19806 19807 80484-800 80484-810 80485-800 80485-810 80485-840
REST19804EA 680 GBP
REST19804 REST19805 REST19806 REST19807 REST80484-800 REST80484-810 REST80485-800 REST80485-810 REST80485-840
GC columns, Rt-XLSulfur (packed and micropacked)
Chromatography Columns
These Rt-XLSulfur columns are optimised for low ppbv sulfur analyses. Interior wall and the end fittings of Rt-XLSulfur columns are Sulfinert treated, making the column as inert as PTFE.
  • Eliminate the need for PTFE tubing
  • Column and end fittings are Sulfinert treated for maximum inertness
  • Maximum temperature of 290 °C
  • Mesh size of 100/120
The packing material for Rt®-XLSulfur columns is extensively deactivated for analysis of low ppbv levels of hydrogen sulfide and methyl mercaptan. It is then treated to achieve effective separation of hydrocarbonsMore Product Information