Porous polymer columns (packed)
Supplier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
80425-800 80425-840 80426-800 80426-810 80426-840 80427-800 80427-810 80427-820 80427-830 80427-840 80433-800 80433-810 80433-840 80467-800 80467-810 80467-840 80468-800 80468-810 80468-840 80469-800 80469-810 80469-820 80470-800 80467-820 80468-820 80470-810
REST80425-800EA 572 GBP
REST80425-800 REST80425-840 REST80426-800 REST80426-810 REST80426-840 REST80427-800 REST80427-810 REST80427-820 REST80427-830 REST80427-840 REST80433-800 REST80433-810 REST80433-840 REST80467-800 REST80467-810 REST80467-840 REST80468-800 REST80468-810 REST80468-840 REST80469-800 REST80469-810 REST80469-820 REST80470-800 REST80467-820 REST80468-820 REST80470-810
Porous polymer columns (packed)
Chromatography Columns
Restek offers a full range of porous polymers, including HayeSep® and Porapak polymer packings for analyses of volatile compounds and light solvents.
  • Siltek®-treated stainless steel
These packed columns are used for the analyses of volatile compounds and light solvents.