XERT-500 column test mix (8 components)
Supplier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
35247
REST35247EA 86 GBP
REST35247
XERT-500 column test mix (8 components)
Standards Chromatography Standards
This contains (C9:0 FAME) methyl nonanoate, (C13) n-tridecane, (C15) n-pentadecane, acenaphthylene, 4-chlorophenol, n-decylamine, 1,6-hexanediol and 1-undecanol (112-42-5).