FAME #12 mix
Supplier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
35021
REST35021EA 164 GBP
REST35021
FAME #12 mix
Standards Food Standards
This mixture can be used for quantification (AOCS method CE 1a-13) and approximate the compositions of oils.
  • Suitable for oils of mid-range to long chain lengths (C13 - C21)
This mix contains five components such as C13 Methyl tridecanoate (20%), C15 methyl pentadecanoate (20%), C17 methyl heptadecanoate (20%), C19 methyl nonadecanoate (20%), C21 methyl heneicosanoate (20%).