FAME #4 mix
Supplier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
35013
REST35013EA 141 GBP
REST35013
FAME #4 mix
Standards Food Standards
This mixture can be used for quantification (AOCS method CE 1a–13) and approximate the compositions of oils.
  • Suitable for oils of mid-range to long chain lengths (C16 - C24)
This mix contains five components such as C16:00 methyl palmitate (20%), C18:00 methyl stearate (20%), C20:00 methyl arachidate (20%), C22:00 methyl behenate (20%), C24:00:00 methyl lignocerate (20%).
Order Now
SPECIFICATIONS
Environmentally Preferable