Steroids and mixed pharmaceuticals mix (10 components)
Supplier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
31117
REST31117EA 119 GBP
REST31117
Steroids and mixed pharmaceuticals mix (10 components)
Standards Environmental Analysis Standards
This steroids and mixed pharmaceuticals mix contain ten components: Bisphenol A, Diclofenac sodium salt, 17-beta-Estradiol, Estrone, 17-alpha-Ethynylestradiol, 4-para-Nonylphenol, 4-tert-Octylphenol, Primidone, Progestrone and Testosterone.
Shelf life 24 months.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.