Pharmaceuticals Mix #1 (8 components)
Supplier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
31116
REST31116EA 112 GBP
REST31116
Pharmaceuticals Mix #1 (8 components)
Standards Organic Standards
This pharmaceuticals mix #1 contain eight components: Acetaminophen, Caffeine, Carbamazepine, Ciprofloxacin HCL, Erythromycin USP, Fluoxetine HCl, Sulphamethoxazole and Trimethoprim.
Shelf life 18 months.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.