NJDEP EPH 10/08 Rev. 2 aliphatics fractionation check mix (18 components)
Supplier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
30544
REST30544EA 286 GBP
REST30544
NJDEP EPH 10/08 Rev. 2 aliphatics fractionation check mix (18 components)
Standards Organic Standards
Consists of 18 components, those are n-nonane, n-decane,  n-dodecane,  n-tetradecane, n-hexadecane, n-octadecane,  n-eicosane,  n-heneicosane,  n-docosane,  n-tetracosane,  n-hexacosane,  n-octacosane,  n-triacontane,  n-dotriacontane,  n-tetratriacontane,  n-hexatriacontane, n-octatriacontane and n-tetracontane.
18 components
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.