ValvTool wrench
Supplier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
25321
REST25321EA 80.5 GBP
REST25321
ValvTool wrench
Wrenches
The ValvTool is a time saving device that provides easy access to many hard-to-reach Rheodyne or Valco valves.