Rethreading tool
Supplier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
21893 23016 23017 23018 23019
REST21893EA 0 GBP
REST21893 REST23016 REST23017 REST23018 REST23019
Rethreading tool
Tool Sets
These rethreading tools are suitable for repairing worn or damaged threads that come as a result of consistent use and temperature changes.
  • Multiple uses (injection ports, fittings, etc.)
  • Built-in guide prevents cross-threading
Repeated installation, removal and temperature changes cause wear and damage to threads on GC parts. This can cause a poor seal, and oxygen can enter the system, compromising analytical results and possibly destroying expensive analytical columns.