Ball valve, Parker Balston® shut-off gas valve
Supplier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
22192 22193 22194 22195
REST22192EA 0 GBP
REST22192 REST22193 REST22194 REST22195
Ball valve, Parker Balston® shut-off gas valve
Valves Vacuum Valves
Ball valves have a floating ball to assist sealing and to reduce operating torque, and dual seats to provide leak-tight bidirectional sealing. They are rated to 1500 psig at 21 °C and have a maximum temperature of about 177 °C.