Protocol wall mounts for gas regulators
Supplier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
21327 21347 21348 21349
REST21327EA 1640 GBP
REST21327 REST21347 REST21348 REST21349
Protocol wall mounts for gas regulators
Wall Hooks
The protocol station is designed for convenient wall mounting of high-purity gas regulators.
  • Ease of use
  • Prevents gas regulator damage and improves safety
Either chrome-plated brass or 316 stainless steel option is complete with a 0,9m, flexible, all-stainless-steel pigtail with armor casing. The CGA connection on the pigtail has an integral check valve in the gland to prevent contamination during cylinder changeout.