Replacement inlet seals
Supplier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
21308 21307 20390 20391 20392 20393 21306 21305 21319 21320 21315 21316 21317 21318
REST21308EA 595 GBP
REST21308 REST21307 REST20390 REST20391 REST20392 REST20393 REST21306 REST21305 REST21319 REST21320 REST21315 REST21316 REST21317 REST21318
Replacement inlet seals
Seals Mechanical Seals
Inlet seals are used at the base of the Agilent 5890/6890/7890 GC injection port, which contacts the sample and should be changed frequently to prevent adsorption of active compounds. In addition, septum fragments and sample residue accumulate on the disk surface, requiring disk replacement.