Inlet liners for Agilent GCs
Supplier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
20795 20797 20798-209.1 20800-209.25 20799-209 20907 21704 20990 20991 20980 20981 20982 20983 20985 20986 20987 20796-214 21704-214 20984 21702 20896-214 20799-210 21055-214 20998-214.25 20954 20974 20975 20989 20997 21052 21053 21703 22408 22409 22410 20771-210.5 20771-214.5 20782-210 20785-210 20785-214 20786-210.25 20786-214.25 20796-200.5 20796-213.5 20797-214.25 20799-214 20799-216 20800-210.25 20800-211.25 20800-214.25 20800-216.25 20908-214 20915-214.5 20981-200.5 20981-213.5 20984-209.5 20984-214 20985-214.25 20989-214 20991-200.5 20997-214.25 21053-214 21054-200.1 21055-200.5 21055-211 21055-213 21390-214.1 21391-214.5 21702-200.5 21702-209 21703-209.25 21704-201.10 21704-210.10 21704-213.10 21704-216 22406-213 22407-213.25 22409-211 20508 20509 22273 24977
REST20795EA 0 GBP
REST20795 REST20797 REST20798-209.1 REST20800-209.25 REST20799-209 REST20907 REST21704 REST20990 REST20991 REST20980 REST20981 REST20982 REST20983 REST20985 REST20986 REST20987 REST20796-214 REST21704-214 REST20984 REST21702 REST20896-214 REST20799-210 REST21055-214 REST20998-214.25 REST20954 REST20974 REST20975 REST20989 REST20997 REST21052 REST21053 REST21703 REST22408 REST22409 REST22410 REST20771-210.5 REST20771-214.5 REST20782-210 REST20785-210 REST20785-214 REST20786-210.25 REST20786-214.25 REST20796-200.5 REST20796-213.5 REST20797-214.25 REST20799-214 REST20799-216 REST20800-210.25 REST20800-211.25 REST20800-214.25 REST20800-216.25 REST20908-214 REST20915-214.5 REST20981-200.5 REST20981-213.5 REST20984-209.5 REST20984-214 REST20985-214.25 REST20989-214 REST20991-200.5 REST20997-214.25 REST21053-214 REST21054-200.1 REST21055-200.5 REST21055-211 REST21055-213 REST21390-214.1 REST21391-214.5 REST21702-200.5 REST21702-209 REST21703-209.25 REST21704-201.10 REST21704-210.10 REST21704-213.10 REST21704-216 REST22406-213 REST22407-213.25 REST22409-211 REST20508 REST20509 REST22273 REST24977
Inlet liners for Agilent GCs
Chromatography Supplies Chromatography Inlet Liners
Inlet liners with deactivation improve trace-level analysis that are suitable for agilent GCs equipped with split/splitless inlets. It is made of borosilicate glass.
  • Increase accuracy and precision by preventing loss of sensitive analytes-even when using wool
  • Lower detection limits for a wide range of active compounds is achieved
  • Liner-to-liner reproducibility through consistent manufacturing and extensive testing