PLOT columns, Rt®-Alumina BOND/MAPD (fused silica)
Supplier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
19780 19779 19781 19777 19778
REST19780EA 2090 GBP
REST19780 REST19779 REST19781 REST19777 REST19778
PLOT columns, Rt®-Alumina BOND/MAPD (fused silica)
Chromatography Columns
Rt-Alumina BOND/MAPD PLOT columns are made specifically for the analysis of petrochemicals and downstream products such as ethylene, propylene, butylenes, and butadiene.
  • Optimised deactivation produces maximum response when analysing trace levels of acetylene, methyl acetylene, and propadiene
  • Stable response factors make this column ideal for process-type applications where recalibration must be minimised
  • High loadability reduces peak tailing and improves separations
  • Extended temperature range up to 250 °C for fast elution of high molecular weight (HMW)
More Product Information