Capillary columns, Rtx®-35 (fused silica)
Supplier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
40402 40410 40411 10420 10421 10423 10424 10435 10437 10438 10439 10440 10453 10454 10455 10470 10485
REST40402EA 0 GBP
REST40402 REST40410 REST40411 REST10420 REST10421 REST10423 REST10424 REST10435 REST10437 REST10438 REST10439 REST10440 REST10453 REST10454 REST10455 REST10470 REST10485
Capillary columns, Rtx®-35 (fused silica)
Chromatography Columns
General-purpose columns for organochlorine pesticides, PCB congeners (e.g. Aroclor mixes), herbicides, pharmaceuticals, sterols, rosin acids, phthalate esters.
Midpolarity phase; Crossbond® diphenyl dimethyl polysiloxane
Temperature range: 40 to 320 °C
Similar phases: HP-35, DB-35, ZB-35.
Equivalent to USP G42 phase.