Sodium thiosulphate 0.05 mol/l (0.05 N) in aqueous solution volumetric solution

Supplier: REAGECON
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate

T20051
REGET20051EA 55.9 GBP
REGET20051
Sodium thiosulphate 0.05 mol/l (0.05 N) in aqueous solution volumetric solution
Sodium thiosulphate
Formula: Na₂S₂O₃
MW: 158.11 g/mol
Boiling Pt: 100
Melting Pt: 48
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR