Sodium thiosulphate 0.01 mol/l (0.01 N) in aqueous solution volumetric solution

Supplier: REAGECON
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

T20011
REGET20011EA 55.9 GBP
REGET20011
Sodium thiosulphate 0.01 mol/l (0.01 N) in aqueous solution volumetric solution
Sodium thiosulphate
Formula: Na₂S₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR