Sodium hydroxide 0.1 mol/l (0.1 N) aqueous solution Volumetric Carbonate_low

Supplier: REAGECON
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Caustic soda

S20101LC
REGES20101LCEA 47.7 GBP
REGES20101LC
Sodium hydroxide 0.1 mol/l (0.1 N) aqueous solution Volumetric Carbonate_low
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 III
Merck Index: 14,08627

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR