Osmolality standards
Supplier: REAGECON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
RE-OSM-500 RE-OSM-50 RE-OSM-100 RE-OSM-200 RE-OSM-400 RE-OSM-850 RE-OSM-1500 RE-OSM-2000 RE-OSM-3000 RE-OSM-900 RE-OSM-1000 RE-OSM-290 RE-OSM-300
REGERE-OSM-500EA 255 GBP
REGERE-OSM-500 REGERE-OSM-50 REGERE-OSM-100 REGERE-OSM-200 REGERE-OSM-400 REGERE-OSM-850 REGERE-OSM-1500 REGERE-OSM-2000 REGERE-OSM-3000 REGERE-OSM-900 REGERE-OSM-1000 REGERE-OSM-290 REGERE-OSM-300
Osmolality standards
Standards Osmolality Standards
Osmolality standards provide ease of use when calibrating all types of osmometers. All Osmolality standards are manufactured using high purity raw materials. These products are prepared gravimetrically on a weight/weight basis. Both solute (salts) and solvent (water) are weighed on a balance calibrated by using OIML traceable weights.
  • Ready to use and presented in convenient ampoules
  • Extended shelf life and has low uncertainty of measurement
  • Consistency of product-Independent, traceable
  • Consist of 12 ampoules of 5 ml
The Osmolality of the standard is verified using a high performance calibrated, temperature controlled osmometer. The calibration of this instrument is completed using high purity certified Osmolality standards.
Certifications: ISO17025 accreditation for calibration of non automatic weighing machines (INAB ref: 265C). The resulting Balance Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.