Potassium iodate 0.05 M aqueous solution

Supplier: REAGECON
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PI2005F
REGEPI2005FEA 273 GBP
REGEPI2005F
Potassium iodate 0.05 M aqueous solution
Potassium iodate
Formula: KlO₃
MW: 214 g/mol
Melting Pt: 560
MDL Number: MFCD00011406
CAS Number: 7758-05-6
Merck Index: 13,07726

Order Now

Specification Test Results

Molarity 0,05 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR