Silver nitrate 0.02 mol/l (0.02 N) in aqueous solution volumetric solution

Supplier: REAGECON
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

N20020W
REGEN20020WEA 966 GBP
REGEN20020W
Silver nitrate 0.02 mol/l (0.02 N) in aqueous solution volumetric solution
Silver nitrate
Formula: AgNO₃
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR