Turbidity standards, low range 20 to 100 NTU
Supplier: REAGECON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CRS-20-500 CRS-100-100 CRS-100-500 CRS-20-100 CRS-50-100 CRS-50-500 CRS-60-500 CRS-60-100 CRSR-100-500 CRSR-20-100 CRSR-20-500 CRSR-40-100 CRSR-40-500 CRSR-50-500 CRSR-90-100 CRSR-90-500 CRSR-50-100 CRSR-100-100 CRS-40-500
REGECRS-20-500EA 524 GBP
REGECRS-20-500 REGECRS-100-100 REGECRS-100-500 REGECRS-20-100 REGECRS-50-100 REGECRS-50-500 REGECRS-60-500 REGECRS-60-100 REGECRSR-100-500 REGECRSR-20-100 REGECRSR-20-500 REGECRSR-40-100 REGECRSR-40-500 REGECRSR-50-500 REGECRSR-90-100 REGECRSR-90-500 REGECRSR-50-100 REGECRSR-100-100 REGECRS-40-500
Turbidity standards, low range 20 to 100 NTU
Standards Turbidity Standards
Turbidity standards for ratio and non-ratio instruments composed of suspended polymer microspheres.
  • Non toxic and non carcinogenic microspheres
  • Highly accurate with well defined spherical shape
  • Traceable to NIST
  • US EPA approved
  • Ready to use- particles stay in suspension