Sodium dihydrogen phosphate

Supplier: G Biosciences
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

RC-098
RC-098EA 167 GBP
RC-098
Sodium dihydrogen phosphate
Sodium dihydrogen phosphate
Formula: NaH₂PO₄
Melting Pt: ∼650 °C
Density: 0.9996 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003527
CAS Number: 7558-80-7
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR