γ-(N-Morpholino)propanesulphonic acid (MOPS)

Supplier: G Biosciences
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid

RC-073 RC-072
RC-073EA 353 GBP
RC-073 RC-072
γ-(N-Morpholino)propanesulphonic acid (MOPS)
γ-(N-Morpholino)propanesulphonic acid (MOPS)
Formula: C₇H₁₅NO₄S
MW: 209.27 g/mol
Melting Pt: > 250 °C
Flash Pt: 110 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00006183
CAS Number: 1132-61-2
EINECS: 214-478-5
Merck Index: 13,06288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR