Microtube refill system for micro-racks, ratiolab®
Supplier: RATIOLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8200005 8200006 8200025 8200026
RATI8200005EA 538 GBP
RATI8200005 RATI8200006 RATI8200025 RATI8200026
Microtube refill system for micro-racks, ratiolab®
Tubes Microtubes
Plastic rack with cardboard box.
  • Available with coded or non-coded Micro-Tubes
  • For the very efficient, hygienic, and fast loading of Micro-Racks
  • Convenient handling and reduced waste
  • Pressure-tested tightness of the Micro-Tubes
The Micro-Tubes are placed on an 8×12 tray and packed in an environmentally friendly cardboard box. The loaded tray can easily be moved from the cardboard box into the empty Micro-Rack.