Tip boxes, Multibox® plus / Multibox® ultra
Supplier: RATIOLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8030160 8030170 8010161
RATI8030160EA 20.1 GBP
RATI8030160 RATI8030170 RATI8010161
Tip boxes, Multibox® plus / Multibox® ultra
Pipette Tip Boxes and Racks
Reusable pipette tip boxes for the ratiolab® MultiTray® refill systems.
  • System simplifies the handling of pipette tips and provides more safety
  • Space and cost saving
  • Reduces the amount of waste
  • Boxes can be autoclaved several times at 121 °C, 20 min
Multibox® plus: Made from PC, with dust-proof hinged and sliding lid, completely removable.
Multibox® ultra: Made from PP, with hinged lid.
MultiTray®, the dust-proof refill packs made of plastics for ratiolab® pipette tips, hold the tips in plastic matrices. Up to five plastic matrices filled with tipsMore Product Information