Serological pipettes
Supplier: RATIOLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3110011 3125011 3150011 3102011
RATI3110011EA 117 GBP
RATI3110011 RATI3125011 RATI3150011 RATI3102011
Serological pipettes
Pipettes Serological Pipettes
Disposable pipettes made from clear polystyrene (PS).
  • Free from RNase, DNase, DNA and pyrogens
  • Sterile, individually packed
  • From one mould – hence more accurate and stable and with minimal residual wetting
  • Permanent black embossed graduations
  • Colour coded cotton wool plug for convenient identification
  • Negative graduation for additional capacity
  • 5, 10 and 25 ml pipettes with scales for dispensed and the residual volumes