ABL90 FLEX PLUS Blood Gas Analyser
Supplier: RADIOMETER CLINICAL PRODUCTS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ABL90FLEXPLUS
RADCABL90FLEXPLUSEA 0 GBP
RADCABL90FLEXPLUS
ABL90 FLEX PLUS Blood Gas Analyser
Clinical Diagnostic Supplies Clinical Analysers
ABL90 FLEX PLUS is the blood gas analyser of choice when the workflow demands quick analysis, efficiency and extensive insights from small sample sizes.
  • Fast analysis
  • Reliable results
  • Easy to use and maintain
Fast analysis of up to 19 parameters:
The ABL90 FLEX PLUS delivers test results in just 35 seconds on up to 19 vital parameters, including creatinine and urea - all from one blood sample of only 65 µl*. With a high uptime of more than 23,5 hours per day and only 60 seconds between each sample measurement, the analyser is quickly ready for you toMore Product Information